Learn More
Aviva Systems Biology Corp MPDZ Antibody - middle region (ARP42895_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42895_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MPDZ. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MPDZ. Alternate names: DKFZp781P216, FLJ25909, FLJ34626, FLJ90240, MUPP1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58354