Learn More
ABclonal Technology MPDZ Rabbit pAb
SDP
Catalog No. 5000106943
Encompass_Preferred
The protein encoded by this gene has multiple PDZ domains, which are hallmarks of protein-protein interactions. The encoded protein is known to interact with the HTR2C receptor and may cause it to clump at the cell surface. Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-06943 Supplier ABclonal Technology Supplier No. A15344500UL
The protein encoded by this gene has multiple PDZ domains, which are hallmarks of protein-protein interactions. The encoded protein is known to interact with the HTR2C receptor and may cause it to clump at the cell surface. Multiple transcript variants encoding different isoforms have been found for this gene.