Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creative Pegworks MPEG-ALKYNE, MW 20K 1G
SDP

Catalog No. NC1766253
Encompass

mPEG-Alkyne, MW 20k, 1g

Catalog No. NC1766253 Supplier Creative Pegworks Supplier No. PSB20321G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

mPEG-Alkyne, MW 20k, 1g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.