Learn More
Medchemexpress LLC Mpeg-amine (MW 1000) | 80506-64-5 | 99.19% | (C2H4O)nC3H9NO | 100 MG
SDP
Catalog No. 5000352930
Encompass_Preferred
m-PEG-NH2 (MW 1000) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates degradation of target proteins
  • Utilizes the ubiquitin-proteasome system
Catalog No. 50-003-52930 Supplier Medchemexpress LLC Supplier No. HY140675100MG
m-PEG-NH2 (MW 1000) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates degradation of target proteins
  • Utilizes the ubiquitin-proteasome system