Learn More
Medchemexpress LLC mPEG-amine (MW 1000) | 80506-64-5 | 99.2% | (C2H4O)nC3H9NO | 500 MG
SDP
Catalog No. 5000355643
Encompass_Preferred
mPEG-amine (MW 1000) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are compounds that connect an E3 ubiquitin ligase ligand with a target protein ligand via a linker. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used for the synthesis of PROTACs
  • Utilizes ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-55643 Supplier Medchemexpress LLC Supplier No. HY140675500MG
mPEG-amine (MW 1000) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are compounds that connect an E3 ubiquitin ligase ligand with a target protein ligand via a linker. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used for the synthesis of PROTACs
  • Utilizes ubiquitin-proteasome system
  • Selectively degrades target proteins