Learn More
Medchemexpress LLC mPEG-amine (MW 10000) | 80506-64-5 | 637448 | 98.0% | (C2H4O)nC3H9NO | 100 MG
SDP
Catalog No. 5000353771
Encompass_Preferred
mPEG-amine (MW 10000), also known as mPEG-NH2 (MW 10000), is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase and the other targets a specific protein, enabling the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables selective protein degradation
  • Average molecular weight of 10000
  • Appears as a white to off-white solid
Catalog No. 50-003-53771 Supplier Medchemexpress LLC Supplier No. HY140678100MG
mPEG-amine (MW 10000), also known as mPEG-NH2 (MW 10000), is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase and the other targets a specific protein, enabling the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables selective protein degradation
  • Average molecular weight of 10000
  • Appears as a white to off-white solid