Learn More
Medchemexpress LLC Mpeg-amine (mw 10000) | 80506-64-5 | 98.0% | (C2H4O)nC3H9NO | 50 MG
SDP
Catalog No. 5000356612
Encompass_Preferred
mPEG-amine (mPEG-NH2) (MW 10000) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-56612 Supplier Medchemexpress LLC Supplier No. HY14067850MG
mPEG-amine (mPEG-NH2) (MW 10000) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins