Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MPEG-amine (MW 2000) | 80506-64-5 | 99.6% | (C2H4O)nC3H9NO | 100 MG
SDP

Catalog No. 5000356613
Encompass_Preferred

MPEG-amine (MW 2000) is a PEG-based PROTAC linker, also known as mPEG-NH2 (MW 2000). It is designed for use in the synthesis of PROTACs. This product is intended for research purposes only.

  • White to off-white solid appearance
  • Purity of 99.57%
  • Soluble in DMSO
  • Recommended for sealed storage at 4°C, away from moisture and light

Catalog No. 50-003-56613 Supplier Medchemexpress LLC Supplier No. HY140676100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MPEG-amine (MW 2000) is a PEG-based PROTAC linker, also known as mPEG-NH2 (MW 2000). It is designed for use in the synthesis of PROTACs. This product is intended for research purposes only.

  • White to off-white solid appearance
  • Purity of 99.57%
  • Soluble in DMSO
  • Recommended for sealed storage at 4°C, away from moisture and light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.