Learn More
Medchemexpress LLC MPEG-amine (MW 20000) | 80506-64-5 | 98.0% | (C2H4O)nC3H9NO | 100 MG
SDP
Catalog No. 5000355468
Encompass_Preferred
m-PEG-NH2 (MW 20000) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Purity: 98.0%
Catalog No. 50-003-55468 Supplier Medchemexpress LLC Supplier No. HY140679100MG
m-PEG-NH2 (MW 20000) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Purity: 98.0%