Learn More
Medchemexpress LLC MPEG-amine (MW 20000) | 80506-64-5 | 98.0% | (C2H4O)nC3H9NO | 250 MG
SDP
Catalog No. 5000352776
Encompass_Preferred
MPEG-amine (MW 20000) is a PEG-based PROTAC linker, crucial for the synthesis of PROTACs. These PROTACs are innovative molecules that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins, featuring two distinct ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other for the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system to degrade target proteins
  • Average molecular weight of 20000
  • Appears as a solid
  • Color is white to off-white
Catalog No. 50-003-52776 Supplier Medchemexpress LLC Supplier No. 250MGHY140679
MPEG-amine (MW 20000) is a PEG-based PROTAC linker, crucial for the synthesis of PROTACs. These PROTACs are innovative molecules that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins, featuring two distinct ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other for the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system to degrade target proteins
  • Average molecular weight of 20000
  • Appears as a solid
  • Color is white to off-white