Learn More
Medchemexpress LLC Mpeg-amine (mw 5000) | 80506-64-5 | 99.6% | (C2H4O)nC3H9NO | 100 MG
SDP
Catalog No. 5000354936
Encompass_Preferred
mPEG-amine (mPEG-NH2) (MW 5000) is a modifier used to increase the water solubility of long-wavelength voltage-sensitive dyes (VSD) or Pittsburgh (PGH) dyes by replacing their sulfonic acid portion. It can also form amide bonds with carboxyl groups on the surface of microspheres under the mediation of EDC and Sulfo-NHS.
  • Increases water solubility of dyes
  • Forms amide bonds with carboxyl groups
  • Suitable for large-scale rotational cytoplasmic flow studies
Catalog No. 50-003-54936 Supplier Medchemexpress LLC Supplier No. HY140677100MG
mPEG-amine (mPEG-NH2) (MW 5000) is a modifier used to increase the water solubility of long-wavelength voltage-sensitive dyes (VSD) or Pittsburgh (PGH) dyes by replacing their sulfonic acid portion. It can also form amide bonds with carboxyl groups on the surface of microspheres under the mediation of EDC and Sulfo-NHS.
  • Increases water solubility of dyes
  • Forms amide bonds with carboxyl groups
  • Suitable for large-scale rotational cytoplasmic flow studies