Learn More
*IMAGION BIOSYSTEMS INC PrecisionMRX mPEG Coated Iron Oxide Nanoparticles-10mg
Catalog No. NC1837705
Encompass
Superparamagnetic iron oxide nanoparticles with a core diameter of 24 – 26 nm and a low size dispersity (≤7%) as measured by Small Angle X-ray Scattering (SAXS). The cores are coated with a monolayer of oleic acid, a monolayer of an amphiphilic polymer, and a monolayer of methoxypolyethylene glycol (mPEG).
Catalog No. NC1837705 Supplier *IMAGION BIOSYSTEMS INC Supplier No. PG010S
Superparamagnetic iron oxide nanoparticles with a core diameter of 24 – 26 nm and a low size dispersity (≤7%) as measured by Small Angle X-ray Scattering (SAXS). The cores are coated with a monolayer of oleic acid, a monolayer of an amphiphilic polymer, and a monolayer of methoxypolyethylene glycol (mPEG).