Learn More
Medchemexpress LLC MPEG-PLA (PEG MW 3000 & PLA MW 50,000) | PEG MW 3000 & PLA MW 50,000 | 100 MG
SDP
Catalog No. 5000348850
Encompass_Preferred
MPEG-PLA (PEG MW 3000 & PLA MW 50,000) is a block copolymer used in laboratory chemical applications and for the manufacture of substances. It can be used to prepare nanoparticles for targeted drug delivery and is intended for research use only.
  • Block copolymer structure
  • Enables preparation of nanoparticles for targeted drug delivery
  • For research use only
Catalog No. 50-003-48850 Supplier Medchemexpress LLC Supplier No. HY139819100MG
MPEG-PLA (PEG MW 3000 & PLA MW 50,000) is a block copolymer used in laboratory chemical applications and for the manufacture of substances. It can be used to prepare nanoparticles for targeted drug delivery and is intended for research use only.
  • Block copolymer structure
  • Enables preparation of nanoparticles for targeted drug delivery
  • For research use only