Learn More
Akina Inc Methoxy Poly(ethylene glycol)-b-Poly(D,L-lactic-co-glycolic) acid copolymer (per g)
SDP
Supplier:  Akina Inc AK1021G
Encompass
mPEG-PLGA (Mw ~5,000:30,000 Da) LA:GA 50:50 Methoxy Poly(ethylene glycol)-b-Poly(D,L-lactic-co-glycolic) acid copolymer (per g)
Catalog No. NC1692688