Learn More
Creative Pegworks mPEG-Silane, MW 550, 5g
SDP
Catalog No. NC3662225
Encompass
mPEG-Silane is a linear monofunctional methoxy PEG with a reactive tri-ethoxy silane group. mPEG-Silane with tri-methoxy is available by custom synthesis. Silane PEGylation is often useful to PEGylate glass and hydroxylated surfaces and particles.
Catalog No. NC3662225 Supplier Creative Pegworks Supplier No. PSB20175G
mPEG-Silane is a linear monofunctional methoxy PEG with a reactive tri-ethoxy silane group. mPEG-Silane with tri-methoxy is available by custom synthesis. Silane PEGylation is often useful to PEGylate glass and hydroxylated surfaces and particles.