Learn More
Sigma Aldrich mPEG5-SH
Catalog No. 503124988 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
500 mg
Catalog No. Quantity
50-312-4988 500 mg
1 options
Catalog No. 50-312-4988 Supplier Sigma Aldrich Supplier No. JKA13012500MG
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Monodisperse PEG. SH can react with metal surfaces, other thiols, disulfide, maleimide and vinyl sulfone groups.Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalizationProduct of JenKem Technology
Specifications
Linear Formula CH3O(CH2CH2O)4 CH2CH2SH
Quantity 500 mg