Learn More
American Radiolabeled Chemicals Inc MPEP-3H 250UC
SDP
Catalog No. 5090104890
Encompass
MPEP-3H 250uCi in ethanol:water (1:1), 1 mCi/ml, S.A. 25-45 Ci/mmol, M.W. 193.24, Exclusive, Shipped in Dry Ice, (2-Methyl-6-(phenylethynl) pyridine)
Catalog No. 50-901-04890 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 1209250UC
Add to Cart
Add to Cart
Request a Quote
MPEP-3H 250uCi in ethanol:water (1:1), 1 mCi/ml, S.A. 25-45 Ci/mmol, M.W. 193.24, Exclusive, Shipped in Dry Ice, (2-Methyl-6-(phenylethynl) pyridine)