Learn More
Medchemexpress LLC MPEP Hydrochloride | 219911-35-0 | 99.82% | 229.70 | 1 ML
SDP
Catalog No. 5000360098
Encompass_Preferred
MPEP Hydrochloride is a potent, selective, noncompetitive, orally active, and systemically active mGlu5 receptor antagonist with an IC50 of 36 nM for completely inhibiting quisqualate-stimulated phosphoinositide (PI) hydrolysis. It also exhibits anxiolytic- or antidepressant-like effects. Additionally, MPEP Hydrochloride serves as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.
  • Potent, selective, noncompetitive mGlu5 receptor antagonist.
  • Orally active and systemically active.
  • Exhibits anxiolytic- or antidepressant-like effects.
  • Functions as a click chemistry reagent due to its Alkyne group, enabling CuAAc reactions with Azide-containing molecules.
Catalog No. 50-003-60098 Supplier Medchemexpress LLC Supplier No. HY1460910MM/1ML
MPEP Hydrochloride is a potent, selective, noncompetitive, orally active, and systemically active mGlu5 receptor antagonist with an IC50 of 36 nM for completely inhibiting quisqualate-stimulated phosphoinositide (PI) hydrolysis. It also exhibits anxiolytic- or antidepressant-like effects. Additionally, MPEP Hydrochloride serves as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.
  • Potent, selective, noncompetitive mGlu5 receptor antagonist.
  • Orally active and systemically active.
  • Exhibits anxiolytic- or antidepressant-like effects.
  • Functions as a click chemistry reagent due to its Alkyne group, enabling CuAAc reactions with Azide-containing molecules.