Learn More
Medchemexpress LLC MPEP (Hydrochloride) | 219911-35-0 | 99.82% | 229.70 | 25 MG
SDP
Catalog No. 5000363354
Encompass_Preferred
MPEP (Hydrochloride) is a potent, selective, noncompetitive, orally active, and systemically active mGlu5 receptor antagonist. It exhibits anxiolytic- or antidepressant-like effects and can be used as a click chemistry reagent.
  • Potent, selective, noncompetitive mGlu5 receptor antagonist
  • Exhibits anxiolytic- or antidepressant-like effects
  • Contains an Alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • IC50 of 36 nM for completely inhibiting quisqualate-stimulated phosphoinositide (PI) hydrolysis
  • Does not show agonist or antagonist activity on human mGlu2, -3, -4a, -7b, -8a, and mGlu6 receptors
Catalog No. 50-003-63354 Supplier Medchemexpress LLC Supplier No. 25MGHY14609
MPEP (Hydrochloride) is a potent, selective, noncompetitive, orally active, and systemically active mGlu5 receptor antagonist. It exhibits anxiolytic- or antidepressant-like effects and can be used as a click chemistry reagent.
  • Potent, selective, noncompetitive mGlu5 receptor antagonist
  • Exhibits anxiolytic- or antidepressant-like effects
  • Contains an Alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • IC50 of 36 nM for completely inhibiting quisqualate-stimulated phosphoinositide (PI) hydrolysis
  • Does not show agonist or antagonist activity on human mGlu2, -3, -4a, -7b, -8a, and mGlu6 receptors