Learn More
Enzo Life Sciences MPEP . hydrochloride (5mg). CAS: 96206-92-7
SDP
Supplier:  Enzo Life Sciences ALX550368M005
Encompass_Preferred
Potent and selective antagonist for mGluR5 receptor. Systemically active in vivo. Alternative name: 6-Methyl-2-(phenylethynyl)-pyridine . HCl. Purity: ≥99%. Appearance: Off-white powder. Solubility: Soluble in water, DMSO or methanol. Long Term Storage: +4°C.
Catalog No. 50-200-9547