Learn More
Cayman Chemical MPEP (hydrochloride), Purity: ≥98%, CAS: 219911-35-0, 50 mg
SDP
Catalog No. 502969392
Encompass_Preferred
A potent, highly selective, and centrally active antagonist at the mGlu5a receptor subtype (IC50 = 36 nM); also has been reported as a positive allosteric modulator of mGluR4 at higher concentrations
Catalog No. 50-296-9392 Supplier Cayman Chemical Supplier No. 1453650 MG
A potent, highly selective, and centrally active antagonist at the mGlu5a receptor subtype (IC50 = 36 nM); also has been reported as a positive allosteric modulator of mGluR4 at higher concentrations