Learn More
Azzota Corp MPF PTFE 25mm 0.22u S 100pk
Catalog No. 5000886940
Encompass_Preferred
Microporous Filter PTFE Diameter 25mm Pore Size 0 22um Sterile 100/pk
Catalog No. 50-008-86940 Supplier Azzota Corp Supplier No. BT250122S
Microporous Filter PTFE Diameter 25mm Pore Size 0 22um Sterile 100/pk