Learn More
Azzota Corp MPF PTFE 47mm 0.45u S 100pk
Catalog No. 5000886990
Encompass_Preferred
Microporous Filter PTFE Diameter 47mm Pore Size 0 45um 100/pk Sterile
Catalog No. 50-008-86990 Supplier Azzota Corp Supplier No. BT470145S
Add to Cart
Add to Cart
Microporous Filter PTFE Diameter 47mm Pore Size 0 45um 100/pk Sterile