Learn More
MilliporeSigma MPGL02GH2 Non Sterile
Supplier:  MilliporeSigma SI1M142A7
Encompass
Device Configuration Capsule | Connections, Inlet/Outlet 6 mm (1/4 in.) Hose Barb with bell | Maximum Differential Pressure, bar (psid) Forward: 4.1 bar (60 psid) @ 25 C; 1.7 bar (25 psid) @ 80 C; 0.35 bar (5 psid) @ 123 C; Reverse: 0.7 bar (10 psid) @ 25 C | Maximum Inlet Pressure, bar (psi) 5.2 bar (75 psi) @ 25 C | Stacked Discs 20 discs
Catalog No. NC1663360