Learn More
ABclonal Technology MPHOSPH6 Rabbit pAb
SDP
Supplier:  ABclonal Technology A157751000UL
Encompass_Preferred
Predicted to enable RNA binding activity. Involved in maturation of 5.8S rRNA. Located in cytosol, nucleolus, and nucleoplasm. Colocalizes with nuclear exosome (RNase complex).
Catalog No. 50-001-05878