Learn More
Cayman Chemical MPI-0441138 10mg
SDP
Catalog No. 502952775
Encompass_Preferred
A potent inducer of apoptosis and growth inhibitor (EC50 = 2 nM for both processes); has excellent blood brain barrier penetration and is effective in mouse xenograft cancer models
Catalog No. 50-295-2775 Supplier Cayman Chemical Supplier No. 1309810 MG
A potent inducer of apoptosis and growth inhibitor (EC50 = 2 nM for both processes); has excellent blood brain barrier penetration and is effective in mouse xenograft cancer models