Learn More
Cayman Chemical mPIPhydrochlorIde 10mg
SDP
Catalog No. 502984592
Encompass_Preferred
A reversible, selective, allosteric antagonist of mGluR7 that blocks agonist-induced calcium mobilization (IC50 = 26 nM)
Catalog No. 50-298-4592 Supplier Cayman Chemical Supplier No. 1886210 MG
A reversible, selective, allosteric antagonist of mGluR7 that blocks agonist-induced calcium mobilization (IC50 = 26 nM)