Learn More
Cayman Chemical MMPIP (hydrochloride), Purity: ≥98%, CAS: 1215566-78-1, 50 mg
SDP
Catalog No. 502984593
Encompass_Preferred
A reversible, selective, allosteric antagonist of mGluR7 that blocks agonist-induced calcium mobilization (IC50 = 26 nM)
Catalog No. 50-298-4593 Supplier Cayman Chemical Supplier No. 1886250 MG
A reversible, selective, allosteric antagonist of mGluR7 that blocks agonist-induced calcium mobilization (IC50 = 26 nM)