Learn More
Enzo Life Sciences Monophosphoryl Lipid A from E. coli, Serotype R515 (Re) (TLRGRADE™) (Ready-to-Use) (1ml)
SDP
Catalog No. 502009582
Encompass_Preferred
Alternative name: MPL-A from E. coli, Serotype R515 (Re). Formulation: Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water. Purity: ≥99.9% (regarding protein or DNA contaminants; KDO not detectable). Long Term Storage: +4°C.
Catalog No. 50-200-9582 Supplier Enzo Life Sciences Supplier No. ALX581203L001
Alternative name: MPL-A from E. coli, Serotype R515 (Re). Formulation: Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water. Purity: ≥99.9% (regarding protein or DNA contaminants; KDO not detectable). Long Term Storage: +4°C.