Learn More
Enzo Life Sciences Monophosphoryl Lipid A from Salmonella minnesota R595 (Re) (TLRGRADE™) (Ready-to-Use) (500µl)
SDP
Catalog No. 502009581
Encompass_Preferred
Adjuvant Alternative name: MPL-A from Salmonella minnesota R595 (Re). Formulation: Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water. Purity: ≥99.9% (regarding protein or DNA contaminants; KDO not detectable). Long Term Storage: +4°C.
Catalog No. 50-200-9581 Supplier Enzo Life Sciences Supplier No. ALX581202R500
Adjuvant Alternative name: MPL-A from Salmonella minnesota R595 (Re). Formulation: Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water. Purity: ≥99.9% (regarding protein or DNA contaminants; KDO not detectable). Long Term Storage: +4°C.