Learn More
Invivogen MPLA-SM Research-Grade TLR4 Agonist, Immunostimulatory, Lyophilized Lipidic Film
SDP
Catalog No. NC2684148
Encompass
InvivoGen MPLA-SM is a research-grade Monophosphoryl Lipid A (MPLA) that acts as a potent TLR4 agonist. It offers the immunostimulatory benefits of LPS without the toxicity associated with native endotoxin. This product is widely used in vaccination studies for its ability to specifically activate TLR4, facilitating the development of effective adjuvants for immunological research. Each lot is rigorously tested to ensure consistent activation of TLR4.
Catalog No. NC2684148 Supplier Invivogen Supplier No. TLRLMPLA
InvivoGen MPLA-SM is a research-grade Monophosphoryl Lipid A (MPLA) that acts as a potent TLR4 agonist. It offers the immunostimulatory benefits of LPS without the toxicity associated with native endotoxin. This product is widely used in vaccination studies for its ability to specifically activate TLR4, facilitating the development of effective adjuvants for immunological research. Each lot is rigorously tested to ensure consistent activation of TLR4.