Learn More
Aviva Systems Biology Corp MPND Antibody - N-terminal region (ARP47698_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47698_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MPND. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MPND. Alternate names: FLJ14981ApplicationWB. Species reactivity: Cow, Dog, Human, Pig
Catalog No. 50-014-59642