Learn More
Apexbio Technology LLC MPO CHLOR/PEROX INHIB KIT-0.1g
SDP
Catalog No. 5000897323
Encompass_Preferred
Myeloperoxidase Chlorination and Peroxidation Inhibitor Screening Assay Kit is a vital tool in biochemical research focusing on the enzyme myeloperoxidase (MPO) which plays a crucial role in the immune response by producing hypochlorous acid from hydrogen peroxide and chloride ions This assay kit helps in identifying inhibitors of MPO s chlorination and peroxidation activities targeting pathways involved in inflammatory conditions and oxidative stress The inhibitors typically exhibit activity in the nanomolar to low micromolar range Common applications include cell model assays animal studies and drug screening processes aimed at developing new anti-inflammatory therapies Experimental concentrations typically depend on the specific research design This kit facilitates the exploration of potential therapeutic compounds enhancing the understanding of MPO s role in various diseases
Catalog No. 50-008-97323 Supplier Apexbio Technology LLC Supplier No. K2314100
Myeloperoxidase Chlorination and Peroxidation Inhibitor Screening Assay Kit is a vital tool in biochemical research focusing on the enzyme myeloperoxidase (MPO) which plays a crucial role in the immune response by producing hypochlorous acid from hydrogen peroxide and chloride ions This assay kit helps in identifying inhibitors of MPO s chlorination and peroxidation activities targeting pathways involved in inflammatory conditions and oxidative stress The inhibitors typically exhibit activity in the nanomolar to low micromolar range Common applications include cell model assays animal studies and drug screening processes aimed at developing new anti-inflammatory therapies Experimental concentrations typically depend on the specific research design This kit facilitates the exploration of potential therapeutic compounds enhancing the understanding of MPO s role in various diseases