Learn More
Cayman Chemical MPO ChlorInatIon Substrate 1ea
SDP
Catalog No. 502896143
Encompass_Preferred
A Clk1 Clk2 and Clk4 inhibitor (IC50s 8 20 and 12 nM respectively) selective for these kinases over Clk3 (IC50 >3000 nM) as well as DYRK1A DYRK1B DYRK2 HIPK1 and HIPK3 (IC50s 213 956 1309 187 and 159 nM respectively) but does inhibit HIPK2 (IC50 23 nM)
Catalog No. 50-289-6143 Supplier Cayman Chemical Supplier No. 7001651 EA
A Clk1 Clk2 and Clk4 inhibitor (IC50s 8 20 and 12 nM respectively) selective for these kinases over Clk3 (IC50 >3000 nM) as well as DYRK1A DYRK1B DYRK2 HIPK1 and HIPK3 (IC50s 213 956 1309 187 and 159 nM respectively) but does inhibit HIPK2 (IC50 23 nM)