Learn More
Cayman Chemical MPO Enzymehumn 1ea
SDP
Catalog No. 502896186
Encompass_Preferred
An activator of the anticipatory unfolded protein response cytotoxic to MCF-7 cells (IC50 42.6 nM) as well as T47D cells at 0.5 and 1 UM
Catalog No. 50-289-6186 Supplier Cayman Chemical Supplier No. 7001661 EA
An activator of the anticipatory unfolded protein response cytotoxic to MCF-7 cells (IC50 42.6 nM) as well as T47D cells at 0.5 and 1 UM