Learn More
Enzo Life Sciences Myeloperoxidase fluorometric detection kit (1x500tests)
SDP
Supplier:  Enzo Life Sciences ADI907029
Encompass_Preferred
The Myeloperoxidase fluorometric detection kit is a one-step no wash homogeneous activity assay that is adaptable to HTS applications. This kit offers highly sensitive fluorescent detection and the ability to monitor multiple time points to follow the kinetics of MPO activity. Alternative Name: MPO. Sensitivity: 0.00195 units/ml. Applications: Fluorescence microscopy. Species Reactivity: Species independent. Contents: Assay buffer concentrate, Detection Reagent, Hydrogen peroxide, Myeloperoxidase enzyme
Catalog No. 50-658-612