Learn More
Medchemexpress LLC HY-115486 10mg Medchemexpress, MPO-IN-28 CAS:37836-90-1 Purity:>98%
SDP
Catalog No. 501969386
Encompass_Preferred
Medchemexpress, HY-115486 10mg MPO-IN-28 CAS:37836-90-1 MPO-IN-28 (Compound 28) is a myeloperoxidase (MPO) inhibitor with an IC50 of 44 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9386 Supplier Medchemexpress LLC Supplier No. HY11548610MG
Medchemexpress, HY-115486 10mg MPO-IN-28 CAS:37836-90-1 MPO-IN-28 (Compound 28) is a myeloperoxidase (MPO) inhibitor with an IC50 of 44 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.