Learn More
Medchemexpress LLC HY-115486 5mg Medchemexpress, MPO-IN-28 CAS:37836-90-1 Purity:>98%
SDP
Catalog No. 501932505
Encompass_Preferred
Medchemexpress, HY-115486 5mg MPO-IN-28 CAS:37836-90-1 MPO-IN-28 (Compound 28) is a myeloperoxidase ( MPO ) inhibitor with an IC 50 of 44 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2505 Supplier Medchemexpress LLC Supplier No. HY1154865MG
Medchemexpress, HY-115486 5mg MPO-IN-28 CAS:37836-90-1 MPO-IN-28 (Compound 28) is a myeloperoxidase ( MPO ) inhibitor with an IC 50 of 44 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.