Learn More
CAPRICO BIOTECHNOLOGIES INC MPO, PerCP-Cyanine5.5, RUO
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4123065
Encompass_Preferred
The MPO PerCP-Cyanine5.5/8/9 RUO antibody from Caprico Biotechnologies is a mouse monoclonal antibody that recognizes myeloperoxidase, an enzyme found in neutrophil granulocytes and monocytes. This antibody is conjugated to PerCP-Cyanine5.5 and is intended for research use only in flow cytometry applications. It has been tested for use in human samples. The antibody is supplied as a liquid in PBS with 0.1% sodium azide and 1% BSA. Each vial contains 100 tests of antibody. This product should be stored at 2-8°C and protected from prolonged exposure to light.
Catalog No. 50-227-9317