Learn More
Apexbio Technology LLC MPP dihydrochloride(Synonyms: MPP dihydrochloride, Methyl-piperidino-pyrazole dihydrochloride, ER-alpha antagonist MPP), 50mg, CAS: 911295-24-4.
SDP
Catalog No. 5000061283
Encompass_Preferred
MPP dihydrochloride (CAS 911295-24-4) is a selective modulator of estrogen receptor (ER) activity demonstrating both agonist and antagonist properties Through modulation of ER-mediated signaling pathways this compound counteracts the biological effects driven by -estradiol inducing apoptosis specifically in ovarian and endometrial cancer cell types Due to its targeted interaction with ER MPP dihydrochloride serves as a research tool for investigating estrogen receptor pathways and their implications in oncology
Catalog No. 50-000-61283 Supplier Apexbio Technology LLC Supplier No. B691050
MPP dihydrochloride (CAS 911295-24-4) is a selective modulator of estrogen receptor (ER) activity demonstrating both agonist and antagonist properties Through modulation of ER-mediated signaling pathways this compound counteracts the biological effects driven by -estradiol inducing apoptosis specifically in ovarian and endometrial cancer cell types Due to its targeted interaction with ER MPP dihydrochloride serves as a research tool for investigating estrogen receptor pathways and their implications in oncology