Learn More
Selleck Chemical LLC MPP dihydrochloride
SDP
Supplier:  Selleck Chemical LLC S8687100MG
Encompass_Preferred
Methyl-piperidino-pyrazole (MPP) dihydrochloride is a highly selective antagonist for estrogen receptor alpha (ER ) with an affinity 200 times greater than for estrogen receptor beta (ER ) It inhibits 17 -estradiol binding to ER by inducing conformational changes that alter the receptor s binding site and also increases the energy required for genistein s interaction with ER
Catalog No. 50-000-00950