Learn More
Selleck Chemical LLC MPP dihydrochloride
Supplier: Selleck Chemical LLC S868725MG
Methyl-piperidino-pyrazole (MPP) dihydrochloride is a highly selective antagonist for estrogen receptor alpha (ER ) with an affinity 200 times greater than for estrogen receptor beta (ER ) It inhibits 17 -estradiol binding to ER by inducing conformational changes that alter the receptor s binding site and also increases the energy required for genistein s interaction with ER