Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mpp dihydrochloride | 911295-24-4 | MFCD06411603 | 99.5% | 542.50 | C29H33Cl2N3O3 | 5 MG
SDP

Catalog No. 5000246687
Encompass_Preferred

MPP dihydrochloride is a selective estrogen receptor (ERα) antagonist provided as a high-purity solid for research use in studies of estrogen signaling. It is used in vitro and in vivo to probe ERα-mediated pathways.

  • High purity: 99.54% (HPLC).
  • Chemical formula: C29H33Cl2N3O3.
  • Molecular weight: 542.50 g/mol.
  • Physical form: solid (white powder).
  • Storage: store sealed at 4°C; in solution: -80°C (6 months), -20°C (1 month).
  • Supplied in small milligram quantities for research use.

Catalog No. 50-002-46687 Supplier Medchemexpress LLC Supplier No. HY1034545MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MPP dihydrochloride is a selective estrogen receptor (ERα) antagonist provided as a high-purity solid for research use in studies of estrogen signaling. It is used in vitro and in vivo to probe ERα-mediated pathways.

  • High purity: 99.54% (HPLC).
  • Chemical formula: C29H33Cl2N3O3.
  • Molecular weight: 542.50 g/mol.
  • Physical form: solid (white powder).
  • Storage: store sealed at 4°C; in solution: -80°C (6 months), -20°C (1 month).
  • Supplied in small milligram quantities for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.