Learn More
Medchemexpress LLC Mpp dihydrochloride | 911295-24-4 | MFCD06411603 | 99.5% | 542.50 g/mol | C29H33Cl2N3O3 | 10 MG

Supplier: Medchemexpress LLC HY10345410MG
MPP dihydrochloride is a research-stage selective estrogen receptor α (ERα) antagonist provided as the dihydrochloride salt for receptor pharmacology and cell-based studies.
- Selective estrogen receptor α (ERα) antagonist.
- Supplied as a solid, high-purity research chemical.
- Cas number 911295-24-4.
- Molecular weight 542.50 g/mol.
- Purity 99.54% (as reported).
- Store at 4°C; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.