Learn More
Medchemexpress LLC MPP dihydrochloride | 911295-24-4 | MFCD06411603 | 99.5% | 542.50 g/mol | C29H33Cl2N3O3 | 50 MG
SDP
Catalog No. 5000244786
Encompass_Preferred
MPP dihydrochloride is a selective estrogen receptor (ER) modulator used in research to probe ER signaling, study agonist/antagonist effects on ERα, and induce apoptosis in estrogen-responsive cancer cell lines. It is supplied as a solid research chemical with high reported purity.
  • Selective ER modulator for receptor pharmacology studies.
  • Induces apoptosis in endometrial and other estrogen-responsive cell lines.
  • High reported purity (≈99.5%).
  • Soluble in DMSO at 30 mg/mL; ultrasonic warming may be required.
  • Store sealed at 4°C; in solution keep at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-44786 Supplier Medchemexpress LLC Supplier No. HY10345450MG
MPP dihydrochloride is a selective estrogen receptor (ER) modulator used in research to probe ER signaling, study agonist/antagonist effects on ERα, and induce apoptosis in estrogen-responsive cancer cell lines. It is supplied as a solid research chemical with high reported purity.
  • Selective ER modulator for receptor pharmacology studies.
  • Induces apoptosis in endometrial and other estrogen-responsive cell lines.
  • High reported purity (≈99.5%).
  • Soluble in DMSO at 30 mg/mL; ultrasonic warming may be required.
  • Store sealed at 4°C; in solution keep at -80°C (6 months) or -20°C (1 month).