Learn More
Medchemexpress LLC MPP dihydrochloride | 911295-24-4 | MFCD20524909 | 99.5% | 542.50 g·mol-1 | C29H33Cl2N3O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10345410MM 1ML
Encompass_Preferred
MPP dihydrochloride is a selective estrogen receptor alpha (ERα) antagonist supplied as the dihydrochloride salt for research use. It is provided as a 10 mM solution in DMSO (1 mL) or as solid quantities for biochemical and cellular studies and shows high reported purity and characterized molecular properties.
  • Selective antagonist of ERα.
  • Available as 10 mM solution in DMSO and multiple solid sizes.
  • High reported purity (≈99.5%).
  • Molecular weight 542.50 g·mol-1.
  • Store sealed, away from moisture; follow cold storage recommendations for solutions.
Catalog No. 50-002-47252