Learn More
Medchemexpress LLC Mpp dihydrochloride | 911295-24-4 | MFCD20524909 | 99.5% | 542.50 g/mol | C29H33Cl2N3O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY103454100MG
Encompass_Preferred
MPP dihydrochloride is a potent, selective estrogen receptor (ER) modulator and antagonist used in research to study ERα-mediated signaling and cell survival pathways. It has documented activity in cellular and in vivo models and is supplied as a high-purity solid or DMSO solution.
  • selective ER modulator with antagonist activity
  • induces apoptosis in endometrial cancer cell lines
  • high purity (99.54%)
  • white to off-white solid; also available as 10 mM in DMSO
  • store sealed, away from moisture; solution stability: -80°C (6 months), -20°C (1 month)
Catalog No. 50-002-44784