Learn More
Medchemexpress LLC HY-W008719 100mg Medchemexpress, MPP+ (iodide) CAS:36913-39-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW008719100MG
Encompass_Preferred
Medchemexpress, HY-W008719 100mg MPP+ (iodide) CAS:36913-39-0 MPP+ iodide, a toxic metabolite of the neurotoxin MPTP, causes symptom of Parkinson's disease in animal models by selectively destroying dopaminergic neurons in substantia nigra. MPP+ iodide is taken up by the dopamine transporter into dopaminergic neurons where it exerts its neurotoxic action on mitochondria by affecting complex I of the respiratory chain. MPP+ iodide is also a high affinity substrate for the serotonin transporter (SERT). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3261