Learn More
Apexbio Technology LLC MPP-IODIDE-10MG
SDP
Catalog No. 5000567526
Encompass_Preferred
MPP iodide (CAS No 36913-39-0) is commonly used in neurotoxicity studies particularly in research on Parkinson s disease It originates as an active metabolite of MPTP which affects the dopaminergic system in the brain The compound targets mitochondrial complex I leading to disruption of oxidative phosphorylation This mechanism results in oxidative stress and neuronal death MPP iodide exhibits activity in the nanomolar to low micromolar range making it a potent tool for in vitro studies It is frequently applied in cell models such as human neuroblastoma cells and in animal models to simulate Parkinsonian symptoms Its role is significant in drug screening and evaluating neuroprotective agents Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67526 Supplier Apexbio Technology LLC Supplier No. C860110
MPP iodide (CAS No 36913-39-0) is commonly used in neurotoxicity studies particularly in research on Parkinson s disease It originates as an active metabolite of MPTP which affects the dopaminergic system in the brain The compound targets mitochondrial complex I leading to disruption of oxidative phosphorylation This mechanism results in oxidative stress and neuronal death MPP iodide exhibits activity in the nanomolar to low micromolar range making it a potent tool for in vitro studies It is frequently applied in cell models such as human neuroblastoma cells and in animal models to simulate Parkinsonian symptoms Its role is significant in drug screening and evaluating neuroprotective agents Experimental concentrations typically depend on the specific research design