Learn More
Medchemexpress LLC Mpp+ iodide | 36913-39-0 | 99.96% | 297.14 | 1 ML
SDP
Catalog No. 5000451171
Encompass_Preferred
MPP+ iodide is a toxic metabolite of the neurotoxin MPTP. It induces symptoms of Parkinson's disease in animal models by selectively damaging dopaminergic neurons in the substantia nigra. It is absorbed by the dopamine transporter into dopaminergic neurons, where it exerts neurotoxic effects on mitochondria by impacting complex I of the respiratory chain. It also serves as a high-affinity substrate for the serotonin transporter (SERT).
  • Selectively damages dopaminergic neurons
  • Exerts neurotoxic effects on mitochondria by impacting complex I of the respiratory chain
  • High-affinity substrate for the serotonin transporter (SERT)
Catalog No. 50-004-51171 Supplier Medchemexpress LLC Supplier No. HYW00871910MM/1ML
MPP+ iodide is a toxic metabolite of the neurotoxin MPTP. It induces symptoms of Parkinson's disease in animal models by selectively damaging dopaminergic neurons in the substantia nigra. It is absorbed by the dopamine transporter into dopaminergic neurons, where it exerts neurotoxic effects on mitochondria by impacting complex I of the respiratory chain. It also serves as a high-affinity substrate for the serotonin transporter (SERT).
  • Selectively damages dopaminergic neurons
  • Exerts neurotoxic effects on mitochondria by impacting complex I of the respiratory chain
  • High-affinity substrate for the serotonin transporter (SERT)